Learn More
Apexbio Technology LLC Palmitoylethanolamide 544-31-0 50mg
SDP
Supplier:  Apexbio Technology LLC B648550
Encompass_Preferred
Palmitoylethanolamide (PEA) is an endogenous lipid mediator structurally related to endocannabinoids but does not directly activate classical cannabinoid receptors It acts primarily as an agonist of the G protein-coupled receptor GPR55 (IC50 approximately 4 M) thereby modulating inflammatory and immune responses In preclinical studies PEA demonstrates potential application in respiratory infection research as a prophylactic by modulating inflammatory pathways and immune responses associated with viral infections Researchers utilize this compound in in vitro and in vivo models to investigate its role in inflammation regulation immune system modulation and related pharmacological pathways involved in respiratory conditions
Catalog No. 50-000-63863