Learn More
Medchemexpress LLC Palonosetron hydrochloride | 135729-62-3 | C19H25ClN2O | 200 MG
SDP
Supplier:  Medchemexpress LLC HYA0021200MG
Encompass_Preferred
Palonosetron hydrochloride is a 5-HT3 antagonist primarily used to prevent acute, delayed, and overall chemotherapy-induced nausea and vomiting. It also exhibits moderate anti-flavivirus activity, potent anti-Zika virus activity in mammalian cells, and possesses antidepressant activity.
  • Prevents acute, delayed, and overall chemotherapy-induced nausea and vomiting
  • Exhibits moderate anti-flavivirus activity
  • Shows potent anti-Zika virus activity in mammalian cells
  • Possesses antidepressant activity
Catalog No. 50-004-32484