Learn More
Medchemexpress LLC Palonosetron hydrochloride (Standard) | 135729-62-3 | C19H25ClN2O | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0021100MG
Encompass_Preferred
Palonosetron hydrochloride is an analytical standard intended for research and analytical applications. This compound acts as a 5-HT3 antagonist, primarily used to prevent acute, delayed, and overall chemotherapy-induced nausea and vomiting. It also demonstrates moderate anti-flavivirus activity, potent anti-Zika virus activity in mammalian cells, and possesses antidepressant activity.
  • Analytical standard for research and analytical applications
  • Functions as a 5-HT3 antagonist
  • Primarily used to prevent chemotherapy-induced nausea and vomiting
  • Shows moderate anti-flavivirus activity
  • Exhibits potent anti-Zika virus activity in mammalian cells
  • Possesses antidepressant activity
  • Suitable for qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS
Catalog No. 50-004-32987