Learn More
Cayman Chemical Palonsetron N-oxIde 25mg
SDP
Supplier:  Cayman Chemical 3421225 MG
Encompass_Preferred
A metabolite of palonosetron; a potential impurity in palonosetron preparations; a degradation product formed by exposure to oxidative stress
Catalog No. 50-294-6646