Learn More
Medchemexpress LLC HY-10655A 5mg Medchemexpress, Palosuran (hydrochloride) CAS:2469274-58-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10655A5MG
Encompass_Preferred
Medchemexpress, HY-10655A 5mg Palosuran (hydrochloride) CAS:2469274-58-4 Palosuran hydrochloride (ACT-058362 hydrochloride) is a potent, selective, and orally active antagonist of urotensin II receptor, with an IC50 of 3.6 nM for CHO cell membranes expressing human recombinant receptors. Palosuran hydrochloride can improves pancreatic and renal function in diabetic rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0692