Learn More
Medchemexpress LLC Palovarotene | 410528-02-8 | 99.5% | 414.54 | 25 MG

Supplier: Medchemexpress LLC 25MGHY14799
Palovarotene is a nuclear retinoic acid receptor γ (RAR-γ) agonist, intended strictly for research use. It presents as an off-white to yellow solid.
- Acts as a nuclear retinoic acid receptor γ (RAR-γ) agonist.
- Suppresses post-traumatic chondrogenesis and osteogenesis, mitigating trauma-induced ectopic bone formation.
- Significantly decreases heterotopic ossification (HO) in mice by 50 to 60%.
- Being investigated in clinical trials for conditions such as Fibrodysplasia Ossificans Progressiva (FOP) and Dry Eye Disease.
- Has a high purity of 99.49%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.