Learn More
Cayman Chemical PAMAM Dendrimer G3 0 Amine w
SDP
Supplier:  Cayman Chemical 39073500 MG
Encompass_Preferred
An amine-terminated PAMAM dendrimer; ~36 Å in diameter; has 32 surface groups; cytotoxic to DU145 cells (IC50 = ~13.42 µM); toxic to zebrafish embryos, inducing mortality 72 hours post-fertilization at 0.2 µM; liposomes coated with PAMAM G3.0 and encapsulating berberine and chrysophanol reduce hydrogen peroxide-induced production of ROS in ARPE-19 cells and protect against retinal damage in a rat model of photooxidation-induced AMD
Catalog No. 50-291-7230