Learn More
Medchemexpress LLC HY-U00175 5mg Medchemexpress, Pamicogrel CAS:101001-34-7 Purity:>98%
Supplier: Medchemexpress LLC HYU001755MG
Medchemexpress, HY-U00175 5mg Pamicogrel CAS:101001-34-7 Pamicogrel (KBT3022) is a cyclooxygenase (COX) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.