Learn More
Medchemexpress LLC Pamiparib | 1446261-44-4 | MFCD30489383 | 100.0% | 298.32 g·mol⁻¹ | C16H15FN4O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10404425MG
Encompass_Preferred
Pamiparib is an orally active, potent, and highly selective inhibitor of poly(ADP-ribose) polymerase (PARP) 1 and 2, commonly used as a research reagent in DNA damage response and oncology studies. It is provided as a high-purity solid suitable for in vitro assays and analytical applications.
  • Potent PARP1 and PARP2 inhibition (IC50 ≈ 0.9 nM and 0.5 nM).
  • High purity suitable for analytical use.
  • Available in small-scale packs for laboratory research.
  • Soluble in common organic solvents for stock solution preparation.
  • Supports studies of DNA repair, synthetic lethality, and antineoplastic mechanisms.
Catalog No. 50-002-41888