Learn More
Selleck Chemical LLC Pamoic acid disodium E2660-100MG
SDP
Supplier:  Selleck Chemical LLC E2660100MG
Encompass_Preferred
Pamoic acid disodium a potent GPR35 agonist with an EC50 value of 79 nM induces GPR35a internalization and activates ERK1/2 with EC50 values of 22 nM and 65 nM respectively also potently recruits -arrestin2 to GPR35 showing an antinociceptive effect
Catalog No. 50-000-40725