Learn More
Carbosynth LLC. PAMP-12 (Human) (0.5 mg vial), 1ea, MW: 1618.9, Molecular formula: C77H119N25O14, CAS# 196305-05-2
SDP
Supplier:  Carbosynth LLC. PAM4339V0.5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Phe-Arg-Lys-Lys-Trp-Asn-Lys-Trp-Ala-Leu-Ser-Arg-NH2 Proadrenomedullin N-terminal 20 Peptide (Human, 9-20), H-FRKKWNKWALSR-NH2, Application: Hypotensive Peptide / Major Endogenous Form of PAMP, Storage Temperature: -20C, References: K. Kuwasato, K. Kitamura, Y. Ishiyama, H. Washimine, J. Kato, K. Kangawa, and T. Eto, FEBS Lett., 414, 105 (1997). (Original)
Catalog No. 50-168-6128