Learn More
Medchemexpress LLC Pamufetinib (TAS-115) | 1190836-34-0 | 98.3% | 518.56 g/mol | C27H23FN4O4S | 10MG

Supplier: Medchemexpress LLC HY1242310MG
Pamufetinib (TAS-115) is an ATP-competitive, small-molecule multi-kinase inhibitor that targets vascular endothelial growth factor receptor (VEGFR) and hepatocyte growth factor receptor (c-Met), used as a research tool to study kinase signaling and tumor biology. Reported inhibitory activity is in the low-nanomolar range, making it suitable for in vitro and in vivo experimental applications.
- Potent dual VEGFR and c-Met inhibition with low-nanomolar IC50s.
- ATP-competitive small-molecule inhibitor suitable for biochemical and cell-based assays.
- High purity (98.3%) appropriate for research applications.
- Molecular weight 518.56 g/mol; formula C27H23FN4O4S.
- Supplied as an off-white to light yellow solid; mesylate form also available.
- Recommended storage: powder at -20°C; solutions at -80°C for long-term storage.
- For research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.