Learn More
ILEX LIFE SCIENCES LLC PAN Biotech L Glutamine Penicillin Streptomycin Mix in saline solution cell culture media supplement 600 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0619100X6
Encompass_Preferred
PAN-Biotech L-Glutamine Penicillin Streptomycin Solution (200 mM L-Glutamine 10 000 U/ml Penicillin 10 mg/ml Streptomycin in 0 9% NaCl) 6 x 100 ml DESCRIPTION Penicillin and streptomycin (Pen-Strep) antibiotics are used to prevent bacterial contamination of cell cultures given their combined action against gram-positive and gram-negative bacteria L-Glutamine an essential amino acid required by all mammalian and insect cells grown in culture serves as an additional source of carbon and energy and in particular is used for nucleic acid synthesis APPLICATION Used to supplement culture media to control bacterial contamination while also supplying energy to cells COMPOSITION 29 320 00 mg/L L-Glutamine 10 000 00 mg/L Streptomycin sulfate 9 000 00 mg/L Sodium chloride 10 000 000 00 Units/L Penicillin G potassium salt STERILE Yes STORAGE -20 C MANUFACTURER PAN-Biotech GmbH ORIGIN Germany USAGE Lab Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80285