Learn More
ILEX LIFE SCIENCES LLC PAN Biotech MEM Eagle Cell Culture Medium with EBSS With L Glutamine and Sodium Pyruvate GMP Grade 500 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC CT08056
Encompass_Preferred
PAN-Biotech MEM Eagle w EBSS w 2 mM L-Glutamine w 1 mM Sodium pyruvate w NEAA w 1 5 g/L NaHCO3 GMP Grade 500 ml DESCRIPTION PAN-Biotech MEM Eagle GMP Grade is a ready-to-use classical basal cell culture medium designed for direct use in GMP-regulated clinical and industrial applications Defined raw materials high batch-to-batch consistency and comprehensive documentation make MEM Eagle GMP a true ready-to-use solution suitable for immediate implementation in development scale-up and manufacturing workflows EBSS Contains Earle s Balanced Salt Solution MEM-NEAA With non-essential amino acids QUALITY GMP Grade (EU Annex 1) BICARBONATE With 1 5 g/L Sodium Bicarbonate GLUCOSE With glucose GLUTAMINE With 2 mM L-Glutamine HEPES Without HEPES PHENOL RED With Phenol Red SODIUM PYRUVATE With 1 mM Sodium Pyruvate STERILE Yes STORAGE 2-8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany
Catalog No. 50-005-80422