Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Nystatin Suspension 10000 U per ml in DPBS cell culture antifungal 600 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0607800X6
Encompass_Preferred
PAN-Biotech Nystatin Suspension 10 000 U/ml in DPBS cell culture antifungal 6 x 100 ml DESCRIPTION PAN-Biotech Nystatin Suspension 10 000 U/ml in DPBS is a fungistatic and fungicidal polyene antibiotic commonly used as a cell culture media supplement to control fungal contamination Its mode of action is by increasing the permeability of the cell membrane of sensitive fungi by binding to sterols mainly ergosterol Its main action is against Candida species Nystatin is produced by Streptomyces noursei and Streptomyces aureus APPLICATION Nystatin solution is often used as a cell culture media supplement to prevent contamination with fungi it has no antibacterial activity LIQUID/POWDER liquid STERILE Yes STORAGE TEMP -20 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80234