Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Penicillin Streptomycin Solution Cell Culture Antibiotic GMP Grade 100 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC CT07110
Encompass_Preferred
PAN-Biotech Penicillin-Streptomycin Solution (10 000 U/ml Penicillin 10 mg/ml Streptomycin) GMP Grade 100 ml DESCRIPTION Penicillin Streptomycin (Pen-Strep) Solution GMP Grade is a ready-to-use antibiotic solution designed for direct use in GMP-regulated clinical and industrial applications The GMP grade version is based on the proven penicillin streptomycin formulation and is manufactured under strictly controlled GMP conditions Defined quality parameters high batch-to-batch consistency and comprehensive documentation make Penicillin Streptomycin GMP a true ready-to-use solution suitable for immediate implementation in sensitive cell culture and manufacturing workflows STREPTOMYCIN SULFATE 10 000 00 mg/L SODIUM CHLORIDE 8 500 00 mg/L PENICILLIN G POTASSIUM SALT 10 000 000 00 Units/L QUALITY GMP Grade (EU Annex 1) LIQUID/POWEDER Liquid STERILE Yes STORAGE -20 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany
Catalog No. 50-005-80300