Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Sodium Bicarbonate Powder cell culture media supplement 100 GR 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0622100
Encompass_Preferred
PAN-Biotech Sodium Bicarbonate Powder (NaHCO3) for Cell Culture 100 g DESCRIPTION PAN-Biotech Sodium Bicarbonate Powder (NaHCO3) is an alkaline agent commonly used as a cell culture media supplement for the maintenance of appropriate pH when cells are incubated with carbon dioxide Additionally sodium bicarbonate provides some nutritional value when culturing mammalian cells CAS NO 144-55-8 LIQUID/POWDER Powder STERILE No STORAGE TEMP Room Temperature (RT) MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures This product may not be used as a pharmaceutical or veterinary drug agricultural product or food additive
Catalog No. 50-005-80167