Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Sodium Chloride Solution 9 g per L for cell culture 500 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0539500
Encompass_Preferred
PAN-Biotech Sodium Chloride Solution 0 9% for Cell Culture 500 ml DESCRIPTION Sodium Chloride Solution 0 9% for Cell Culture is sterile filtered through a 0 2 m filter Before final release the reagent undergoes extensive quality tests (e g sterility pH value osmolality) COMPOSITION 9 0 g/L sodium chloride in cell culture grade water APPLICATION Sodium chloride (NaCl) solution is used in a wide variety of biochemical applications including density gradients or as diluents to increase ionic strength in buffers or cell culture media It is also important in media to maintain osmotic pressure and/or make it more selective It can also be used as a washing solution for living cells or as a carrier solution for various laboratory substances PRODUCT CATEGORY Media supplements STERILE Yes STORAGE TEMP 2 C - RT MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80264