Learn More
Apexbio Technology LLC Pan-RAS-IN-1 1835283-94-7 1mL(10 mM in DMSO)
SDP
Supplier:  Apexbio Technology LLC BA57675.1
Encompass_Preferred
Pan-RAS-IN-1 (CAS 1835283-94-7) is a small-molecule inhibitor targeting the RAS family of GTPases including KRAS HRAS and NRAS It acts by disrupting the interaction between RAS proteins and their downstream effectors thereby inhibiting RAS-mediated signaling pathways responsible for regulating cell proliferation and oncogenic transformation Pan-RAS-IN-1 is suitable for investigating the functional roles of RAS-dependent signaling in various cellular contexts and is a valuable tool for studies focused on RAS-driven tumorigenesis and the development of targeted therapeutic strategies
Catalog No. 50-000-88790