Learn More
Sigma Aldrich Fine Chemicals Biosciences Pancuronium bromide100MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P1918100MG
Encompass
Pancuronium bromide is a biquarternary amino steroid. It blocks muscarinic acetylcholine receptors (M2) in the heart. Pancuronium has vagolytic and sympathomimetic properties. It is associated with tachycardia and hypertension.
Catalog No. 50-290-2489