Learn More
Apexbio Technology LLC Pancuronium dibromide, 25mg. Cas: 15500-66-0 MFCD: MFCD00079223. AChR antagonist
SDP
Supplier:  Apexbio Technology LLC B161225
Encompass_Preferred
Pancuronium dibromide is a competitive AChR antagonist (IC50 = 5.5 nM) and acts as a skeletal muscle relaxant. Pancuronium dibromide interrupts neuromuscular transmission by competing with acetylcholine for receptor sites on the motor end-plate. Pancuroni Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5274