Learn More
Apexbio Technology LLC Pancuronium dibromide, 15500-66-0, MFCD00079223, 50 mg
SDP
Supplier:  Apexbio Technology LLC B161250
Encompass_Preferred
MF: C35H60N2O42Br, MW: 732.67, Solubility: 36.6335mg/mL in DMSO. Pancuronium dibromide is a competitive AChR antagonist (IC50 = 5.5 nM) and acts as a skeletal muscle relaxant. Pancuronium dibromide interrupts neuromuscular transmission by competing with acetylcholine for receptor sites on the motor end-plate.
Catalog No. 50-101-0641