Learn More
Abcam Pancuronium dibromide, Competitive nicotinic acetylcholine receptor antagonist, 50MG
Supplier:  Abcam AB12053550MG
Encompass_Preferred
MW 732.7 Da. Competitive nicotinic acetylcholine receptor (nAChR) antagonist. Displaces acetylcholine (ab120469) from nAChRs (IC₅₀ = 5.5 nM). Neuromuscular blocker and muscle relaxant.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7510