Learn More
Hach Company Panel Mount Kit for SC1000
Supplier:  Hach Company 6169900
Encompass
Panel Mount Kit for SC1000 Digital ControllerThe panel mount kit for the SC1000 includes a 7 7m extension cable (5796000) panel mount bracket (6168100) and cable strain relief (6128701 5H1017 and 7123500)
Catalog No. 50-002-07237