Learn More
Selleck Chemical LLC Panobinostat (LBH589) 10mM/1mL 404950-80-7 NVP-LBH589
SDP
Supplier:  Selleck Chemical LLC S103010MM/1ML
Encompass_Preferred
Panobinostat (LBH589, NVP-LBH589) is a novel broad-spectrum HDAC inhibitor with IC50 of 5 nM in a cell-free assay. Panobinostat (LBH589) induces autophagy and apoptosis. Panobinostat effectively disrupts HIV latency in vivo. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1921159