Learn More
Selleck Chemical LLC Pantoprazole S2105-100mg
SDP
Supplier:  Selleck Chemical LLC S2105100MG
Encompass_Preferred
Pantoprazole (SKF96022 BY-1023) is a proton pump inhibitor drug that inhibits gastric acid secretion It works on gastric parietal cells to irreversibly inhibit (H /K )-ATPase function and suppress the production of gastric acid
Catalog No. 50-311-0151