Learn More
Medchemexpress LLC Pantoprazole (sodium) | 138786-67-1 | 99.8% | 405.35 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY17507A10MM/1ML
Encompass_Preferred
Pantoprazole sodium (BY1023 sodium) is an orally active and potent proton pump inhibitor (PPI). This substituted benzimidazole acts as a strong H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. It enhances pH stability and exhibits anti-secretory and anti-ulcer properties. When combined with Doxorubicin, Pantoprazole sodium significantly increased tumor growth delay.
  • Potent H+/K+-ATPase inhibitor
  • Improved pH stability
  • Anti-secretory activity
  • Anti-ulcer activity
  • Potential in cancer therapy
  • Blocks exosome release
  • Impairs tumor cell acidification
Catalog No. 50-003-86215