Learn More
Medchemexpress LLC Pantoprazole (sodium) | 138786-67-1 | 99.8% | 405.35 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17507AR
Encompass_Preferred
Pantoprazole sodium is an orally active and potent proton pump inhibitor (PPI). This substituted benzimidazole acts as a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. It improves pH stability and exhibits anti-secretory and anti-ulcer activities. Additionally, it has been shown to significantly increase tumor growth delay when combined with Doxorubicin.
  • Orally active and potent proton pump inhibitor.
  • Potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM.
  • Improves pH stability.
  • Possesses anti-secretory and anti-ulcer activities.
  • Increases tumor growth delay in combination with Doxorubicin.
Catalog No. 50-003-84386