Learn More
Medchemexpress LLC Pantoprazole sodium hydrate | 164579-32-2 | 99.93% | 432.37 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY17507B
Encompass_Preferred
Pantoprazole sodium hydrate is an orally active and potent proton pump inhibitor (PPI) and a substituted benzimidazole. It is a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. It improves pH stability and possesses anti-secretory and anti-ulcer activities. It also significantly increased tumor growth delay when combined with Doxorubicin.
  • Orally active and potent proton pump inhibitor
  • Substituted benzimidazole
  • Potent H+/K-ATPase inhibitor with an IC50 of 6.8 μM
  • Improves pH stability
  • Anti-secretory and anti-ulcer activities
  • Increases tumor growth delay in combination with Doxorubicin
Catalog No. 50-003-85459