Learn More
Hach Company Phenylarsine Oxide (PAO) Standard Solution, 0.00564 N, 1 L
Supplier:  Hach Company 199953
Encompass
For chlorine determination by amperometric titration methods. Note: also available in Digital Titrator cartridges. 0.00564 N, 1 liter bottle
Catalog No. 50-199-4656