Learn More
Hach Company Phenylarsine Oxide (PAO) Standard Solution, 0.00564 N, 3.78 L
Supplier:  Hach Company 199917
Encompass
For chlorine determination by buret titration. Phenylarsine Oxide (PAO) Standard Solution, 0.00564. 3.78 L Note: also available in Digital Titrator cartridges.
Catalog No. 50-199-4654