Learn More
Medchemexpress LLC HY-10015 5mg Medchemexpress, PAP-1 CAS:870653-45-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100155MG
Encompass_Preferred
Medchemexpress, HY-10015 5mg PAP-1 CAS:870653-45-5 PAP-1 (5-(4-Phenoxybutoxy)psoralen) is a potent, selective, and orally active Kv1.3 blocker ( EC 50 =2 nM). PAP-1 blocks Kv1.3 in a use-dependent manner and acts by preferentially binding to the C-type inactivated state of the channel. PAP-1 exhibits 23-fold selectivity over Kv1.5 ( EC 50 =45 nM), and further displays 33- to 125-fold selectivity over all other Kv1-family channels. PAP-1 does not exhibit cytotoxic or phototoxic effects [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1732