Learn More
Genetex Inc Placental Alkaline Phosphatase Monoclonal Antibody [MSVA-350R] HistoMAX™ (GTX04413) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04413
Encompass_Preferred
The Placental Alkaline Phosphatase antibody [MSVA-350R] HistoMAX™ (GTX04413) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human placenta (mature) tissue using GTX04413 Placental Alkaline Phosphatase antibody [MSVA-350R] HistoMAX. Placental Alkaline Phosphatase immunostaining increases continuously throughout the pregnancy. Placental Alkaline Phosphatase immunostaining is strong in the cyto- and syncytiotrophoblast of the mature placenta.
Catalog No. NC3666872