Learn More
Genetex Inc Prostatic Acid Phosphatase Monoclonal Antibody [MSVA-452M] HistoMAX™ (GTX04396) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04396
Encompass_Preferred
The Prostatic Acid Phosphatase antibody [MSVA-452M] HistoMAX™ (GTX04396) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human prostate tissue using GTX04396 Prostatic Acid Phosphatase antibody [MSVA-452M] HistoMAX. Strong prostatic acid phosphatase immunostaining in all acinar epithelial cells of the prostate.
Catalog No. NC3666759