Learn More
Sigma Aldrich Fine Chemicals Biosciences PAP pen for immunostaining, 2 mm tip
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6725481EA
Encompass
Use pen to draw a hydrophobic circle around slide-mounted tissue. Prevents the waste of valuable reagents by keeping liquid pooled in a small droplet. Insoluble in ethanol, acetone and water, but will wash off with xylene.
Catalog No. NC0525776