Learn More
Worthington Biochemical Corporation PAPAIN DISSOCIATION SYSTEM PDS
SDP
Supplier:  Worthington Biochemical Corporation LK003153
Encompass
Set of five single use vials of papain and five single use vials of DNase, 100 ml of Earle's balanced salt solution (EBSS), and an inhibitor vial for use in the tissue dissociation method of Huettner, J.E., and Baughman, R.W.: J. Neuroscience
Catalog No. NC9809987