Learn More
Medchemexpress LLC Papain inhibitor (Glycylglycyl-L-tyrosyl-L-arginine) | 70195-20-9 | 99.6% | 451.48 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP1315100MG
Encompass_Preferred
Papain inhibitor (Glycylglycyl-L-tyrosyl-L-arginine) acts as an effective competitive inhibitor to papain, with a Ki of 9 μM at pH 6.2. This high-purity chemical is suitable for research use.
  • Effective competitive inhibitor to papain.
  • Ki of 9 μM at pH 6.2.
  • Available in various quantities (10 mg, 25 mg, 50 mg, 100 mg).
  • High purity (99.60%).
  • For research use only.
Catalog No. 50-004-23037