Learn More
Worthington Biochemical Corporation PAPAIN SUSPENSION 1GM
SDP
Supplier:  Worthington Biochemical Corporation LS003127
Encompass
Supplied as a 2X crystalline suspension in 50 mM sodium acetate, pH 4.5. To insure full activity, the enzyme should be incubated in a solution containing 1.1 mM EDTA, 0.067 mM mercaptoethanol and 5.5 mM cysteine-HCl for 30 minutes. It is recommended that the enzyme be 0.22 micron filtered after dissolution and prior to use. 9001-73-4
Catalog No. 50-592-333