Learn More
AdipoGen Papaverine . hydrochloride,Chemical. CAS: 61-25-6.
SDP
Supplier:  AdipoGen AGCN20414G001
Encompass_Preferred
Papaverine . hydrochloride, AdipoGen Life Sciences. Chemical. CAS: 61-25-6. Formula: C20H21NO4 . HCl. MW: 339.4 . 36.5. Synthetic. Originally isolated from Papaver somniferum. Smooth muscle relaxant and vasodilator. Phosphodiesterase PDE10A inhibitor. Antiviral. Antipsychotic. Potential inducer of browning in WAT.
Catalog No. 50-596-391