Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Crystal PAPER INSERTS 11.0 MM PK/200

Supplier:  International Crystal 00122482

Encompass_Preferred

Paper Inserts With 11.0 mm Dia. Aperture -pk/200

Catalog No. NC3234614


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.