Learn More
Essendant PAPER17X50HOT PRESSNT
Supplier:  Essendant EPSS042323
Encompass_Preferred
100pct cotton rag media with a natural smooth finish Extremely high color gamut and black density 90 ISO Brightness 98pct opacity Acid-free and pH buffered Sheet/Roll Size- 17x50 ft Paper/Media Thickness (mil)- 16 mil Paper/Media Color(s)- Natural Color Family- White
Catalog No. 50-000-35428