Learn More
Essendant PAPERADHESIVE 44X100
Supplier:  Essendant EPSS041619
Encompass_Preferred
Adhesive Synthetic Paper is an ideal choice for signage that requires adhesive backing An affordable polypropylene-base media it s thin light and very flexible with a durable wet-strength base It delivers accurate colors and vivid results on its high-quality matte surface An instant-drying paper it s excellent for indoor banners When laminated it s perfect for outdoor signage applications as well Best of all its adhesive backing makes it possible to remove and reposition signage for easy mounting Sheet/Roll Size- 44x100 ft Paper Grade (gsm)- 135 gsm Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-34279