Learn More
Essendant PAPERCOLD PRESS NT
Supplier:  Essendant EPSS042300
Encompass
The Epson Cold Press Natural Paper is an acid free ph buffered 100pct cotton rag media with a natural textured matte finish This paper has an extremely high color gamut and black density and is designed to deliver the best quality when used with Epson ink Sheet/Roll Size- 13x19 Paper/Media Thickness (mil)- 21 mil Paper/Media Color(s)- Natural Color Family- White
Catalog No. 50-000-32467