Learn More
Essendant PAPERDS TRANSFER44X500
Supplier:  Essendant EPSS450393
Encompass_Preferred
Unique chemical coating technology allows for heavy ink loads with low cockling superior clarity and sharpness that only Epson can deliver For about the same cost as typical branded or other high-quality paper solutions you can have a genuine Epson media technology with full support Textile-adhesive is recommended for high-end fabric and apparel production Production is a lightweight paper ideal for roll-to-roll applications Sheet/Roll Size- 44x500 ft Paper Grade (gsm)- 81 gsm Paper/Media Thickness (mil)- 4 1 mil Paper/Media Color(s)- White
Catalog No. 50-000-35252