Learn More
Essendant PAPERDSLPY TRANS24X100
Supplier:  Essendant EPSS450191
Encompass
This new and improved polyester backlit film features a glare-reducing matte surface and a microporous coating specially formulated to produce rich colors and a high Dmax with Epson pigmented inks Hot or cold lamination can extend the durability and print life of this film Engineered for creating backlit signage to be used in a lightbox State-of-the-art opacifying layer diffuses light throughout the printed image so that it is evenly lit Blacks look deep and dark colors are vibrant and flesh tones are warm Sheet/Roll Size- 24x100 ft Paper/Media Thickness (mil)- 5 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-34995