Learn More
Essendant PAPERENHNCED MTTE17X100
Supplier:  Essendant EPSS450439
Encompass_Preferred
The roll of Enhanced Matte Paper from Epson is a 192 g/m2 bright white paper with a matte finish The paper yields highly saturated images while maintaining highlight and shadow detail Enhanced Matte is designed for use with UltraChrome Epson Archival DURABrite as well as dye-based ink The heavy weight stock helps deliver the true photographic feels Sheet/Roll Size- 17x100 ft Paper/Media Thickness (mil)- 10 3 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-35571