Learn More
Essendant PAPERFINE ART 44X50WE
Supplier:  Essendant EPSS041783
Encompass
Artists and professional photographers require true archival print media in a variety of weights and surface textures for their digital fine art and photography Now Epson brings you an acid free 100pct cotton hot press paper that is coated on both sides for extra versatility This cylinder mould-made product features an ultra-smooth finish on two sides (sheets) that is optimized for Epson Photographic Dye Epson UltraChrome and Archival Inks Offered in a variety of sizes and formats this fine art paper is not only acid lignin and chlorine free it is also pH buffered with calcium carbonate for a true archival sheet Sheet/Roll Size- 44x50 ft Paper Grade (gsm)- 250 gsm Paper/Media Thickness (mil)- 15 mil Paper/Media Color(s)- White
Catalog No. 50-000-34313