Learn More
Essendant PAPERHOT PRESS BRIGHT
Supplier:  Essendant EPSS042327
Encompass
For fine art photographers looking for a matte medium that can deliver a wide color gamut and high brightness Epson has developed Hot Press Bright Paper Made from 100pct cotton rag and using a hot press production technique this paper features a smooth matte finish The heavy weight of 330 gsm thickness of 18 mil and opacity of 98pct further give this paper the look and feel of a true fine art paper Also through the use of optical brightening agents this media will help deliver bright whites deep blacks and a wide color gamut Designed for longevity Hot Press Bright is free of acid lignin and chlorine and is pH buffered Sheet/Roll Size- 8 5x11 Paper/Media Thickness (mil)- 17 mil Paper/Media Color(s)- Bright White Color Family- White
Catalog No. 50-000-35457