Learn More
Essendant PAPERINK PHTO 24X100GLS
Supplier:  Essendant HEWQ8916A
Encompass_Preferred
Microporous surface allows for maximum ink absorption resulting in vibrant colors fast dry time and superior fade resistance Expand your range of photo offerings with this versatile everyday photo paper Instant dry time allows for immediate post-print processing robust handling and lamination Moderately water resistant with UV pigment inks Compatible across various HP and non-HP pigment printers Quick Dry Paper Weight- 235 g/m2 Sheet/Roll Size- 24x100 ft Paper/Media Thickness (mil)- 9 1 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-35205