Learn More
Essendant PAPERMT 36X131
Supplier:  Essendant EPSS041854
Encompass_Preferred
100pct cotton hot press paper with a coating on both sides for extra versatility Features an ultra smooth finish on two sides that is optimized for Epson Photographic Dye Epson UltraChrome and Archival Inks Acid- lignin- and chlorine-free it is also pH buffered with calcium carbonate for a true archival sheet Engineered to give you the highest resolution and color saturation possible Sheet/Roll Size- 36x131 7 ft Paper/Media Thickness (mil)- 5 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-34281